Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zdhhc12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017307.2 | NP_001017307.1 | 2 - 3 | 296 | 81 | |
XM_012970476.2 | XP_012825930.1 | 2 - 3 | 456 | 81 | ||
GenBank mRNA | BC121517.1 | - | 2 - 3 | 595 | 81 | |
BC157776.1 | - | 2 - 3 | 559 | 81 | ||
BC170884.1 | - | 2 - 3 | 275 | 81 | ||
BC170886.1 | - | 2 - 3 | 275 | 81 | ||
CR855529.2 | - | 2 - 3 | 296 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |