Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mmp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030330.1 | NP_001025501.1 | 3 - 4 | 531 | 101 | |
GenBank mRNA | BC080175.1 | - | 3 - 4 | 537 | 101 | |
BC161475.1 | - | 3 - 4 | 524 | 101 | ||
CR760257.1 | - | 3 - 4 | 531 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |