Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hdac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030393.1 | NP_001025564.1 | 8 - 9 | 873 | 110 | |
GenBank mRNA | BC090604.1 | - | 8 - 9 | 873 | 110 | |
CT030593.1 | - | 8 - 9 | 851 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |