Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pdgfrl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030415.2 | NP_001025586.1 | 4 - 5 | 828 | 82 | |
GenBank mRNA | BC091030.1 | - | 4 - 5 | 829 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |