Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pdgfrl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030415.2 | NP_001025586.1 | 5 - 6 | 967 | 89 | |
GenBank mRNA | BC091030.1 | - | 5 - 6 | 968 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |