Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ccdc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030419.1 | NP_001025590.1 | 1 - 2 | 792 | 107 | |
GenBank mRNA | BC091039.1 | - | 1 - 2 | 792 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |