Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prdx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030437.1 | NP_001025608.1 | 423 | 115 | ||
GenBank mRNA | BC091062.1 | - | 423 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Xenopus tropicalis |