Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hao2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030453.1 | NP_001025624.1 | 4 - 5 | 617 | 92 | |
XM_012957078.2 | XP_012812532.1 | 5 - 6 | 816 | 92 | ||
XM_012957079.2 | XP_012812533.1 | 5 - 6 | 813 | 92 | ||
XM_012957080.2 | XP_012812534.1 | 4 - 5 | 687 | 92 | ||
XM_012957081.2 | XP_012812535.1 | 5 - 6 | 885 | 92 | ||
XM_012957082.2 | XP_012812536.1 | 4 - 5 | 1108 | 92 | ||
XM_012957084.1 | XP_012812538.1 | 4 - 5 | 709 | 92 | ||
XM_012957085.2 | XP_012812539.1 | 4 - 5 | 1105 | 92 | ||
XM_012957086.2 | XP_012812540.1 | 4 - 5 | 1149 | 92 | ||
XM_012957087.2 | XP_012812541.1 | 4 - 5 | 761 | 92 | ||
GenBank mRNA | BC091092.1 | - | 4 - 5 | 617 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |