Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pglyrp1-like.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030455.1 | NP_001025626.1 | 1 - 2 | 83 | 78 | |
XM_012968516.2 | XP_012823970.1 | 3 - 4 | 272 | 78 | ||
GenBank mRNA | BC091103.1 | - | 1 - 2 | 83 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |