Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tpst1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030469.1 | NP_001025640.1 | 3 - 4 | 1214 | 86 | |
XM_012957093.2 | XP_012812547.2 | 3 - 4 | 1273 | 86 | ||
GenBank mRNA | BC091606.1 | - | 3 - 4 | 1214 | 86 | |
CR942619.2 | - | 3 - 4 | 1193 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |