Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gabarapl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030481.1 | NP_001025652.1 | 3 - 4 | 372 | 71 | |
GenBank mRNA | BC091701.1 | - | 3 - 4 | 372 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |