Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mettl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030492.1 | NP_001025663.1 | 2 - 3 | 166 | 92 | |
GenBank mRNA | BC092545.1 | - | 2 - 3 | 166 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |