Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mrgbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030510.1 | NP_001025681.1 | 181 | 58 | ||
GenBank mRNA | BC093470.1 | - | 181 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis |