Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pex11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030530.1 | NP_001025701.1 | 2 - 3 | 257 | 62 | |
XM_012959137.1 | XP_012814591.1 | 3 - 4 | 378 | 62 | ||
GenBank mRNA | BC090557.1 | - | 2 - 3 | 257 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |