Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vangl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032310.1 | NP_001027481.1 | 4 - 5 | 794 | 99 | |
XM_012969845.1 | XP_012825299.1 | 2 - 3 | 398 | 99 | ||
GenBank mRNA | BC095909.1 | - | 4 - 5 | 794 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |