Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slco1b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032322.1 | NP_001027493.1 | 7 - 8 | 1551 | 66 | |
GenBank mRNA | BC096506.1 | - | 7 - 8 | 1551 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |