Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: atp2a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032327.1 | NP_001027498.1 | 18 - 19 | 2794 | 78 | |
XM_012954931.2 | XP_012810385.1 | 18 - 19 | 2826 | 78 | ||
GenBank mRNA | BC096519.1 | - | 18 - 19 | 2794 | 78 |
Gene Symbol: LOC101733483 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018090443.1 | XP_017945932.1 | 10 - 11 | 1995 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |