Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032329.1 | NP_001027500.1 | 3 - 4 | 405 | 86 | |
XM_012966676.2 | XP_012822130.1 | 3 - 4 | 833 | 86 | ||
GenBank mRNA | BC096632.1 | - | 3 - 4 | 405 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |