Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032329.1 | NP_001027500.1 | 4 - 5 | 610 | 100 | |
XM_012966676.2 | XP_012822130.1 | 4 - 5 | 1038 | 100 | ||
GenBank mRNA | BC096632.1 | - | 4 - 5 | 610 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |