Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032329.1 | NP_001027500.1 | 5 - 6 | 850 | 75 | |
XM_012966676.2 | XP_012822130.1 | 5 - 6 | 1278 | 75 | ||
GenBank mRNA | BC096632.1 | - | 5 - 6 | 850 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |