Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bcl2l11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032340.1 | NP_001027511.1 | 3 - 4 | 746 | 80 | |
XM_012963081.2 | XP_012818535.1 | 3 - 4 | 2056 | 80 | ||
GenBank mRNA | BC098324.1 | - | 3 - 4 | 746 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |