Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: xkr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033942.2 | NP_001029114.1 | 5 - 6 | 895 | 90 | |
XM_012964043.2 | XP_012819497.1 | 5 - 6 | 930 | 90 | ||
XM_018089354.1 | XP_017944843.1 | 4 - 5 | 775 | 90 | ||
GenBank mRNA | AY699708.1 | - | 5 - 6 | 898 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Xenopus tropicalis Probe spans exons |