Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ddi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033948.1 | NP_001029120.1 | 4 - 5 | 807 | 59 | |
GenBank mRNA | BC100609.1 | - | 4 - 5 | 807 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |