Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pdcd2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007507.3 | NP_001007508.2 | 5 - 6 | 787 | 81 | |
XM_012960904.2 | XP_012816358.1 | 5 - 6 | 802 | 81 | ||
XM_012960906.2 | XP_012816360.1 | 3 - 4 | 440 | 81 | ||
XM_018093011.1 | XP_017948500.1 | 5 - 6 | 821 | 81 | ||
XM_018093012.1 | XP_017948501.1 | 5 - 6 | 821 | 81 | ||
GenBank mRNA | BC079944.1 | - | 5 - 6 | 787 | 81 | |
CR848635.2 | - | 3 - 4 | 401 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |