Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hey1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007910.2 | NP_001007911.1 | 1 - 2 | 243 | 62 | |
GenBank mRNA | BC080349.1 | - | 1 - 2 | 203 | 62 | |
CR760684.2 | - | 1 - 2 | 243 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |