Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: traf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008161.2 | NP_001008162.2 | 5 - 6 | 913 | 92 | |
GenBank mRNA | BC082342.1 | - | 5 - 6 | 915 | 92 | |
CR855449.2 | - | 5 - 6 | 913 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |