Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: traf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008161.2 | NP_001008162.2 | 6 - 7 | 998 | 63 | |
GenBank mRNA | BC082342.1 | - | 6 - 7 | 1000 | 63 | |
CR855449.2 | - | 6 - 7 | 998 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |