Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: golph3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011166.2 | NP_001011166.1 | 2 - 3 | 243 | 77 | |
XM_012969831.2 | XP_012825285.1 | 2 - 3 | 302 | 77 | ||
GenBank mRNA | BC084991.1 | - | 2 - 3 | 219 | 77 | |
CR761980.1 | - | 2 - 3 | 207 | 77 | ||
CR848444.2 | - | 2 - 3 | 243 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |