Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tm2d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011324.2 | NP_001011324.1 | 2 - 3 | 422 | 78 | |
GenBank mRNA | BC088075.1 | - | 2 - 3 | 308 | 78 | |
CR761327.2 | - | 2 - 3 | 422 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |