Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cpeb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015925.2 | NP_001015925.1 | 7 - 8 | 1653 | 65 | |
XM_012966552.2 | XP_012822006.1 | 6 - 7 | 1863 | 65 | ||
XM_018095384.1 | XP_017950873.1 | 8 - 9 | 1764 | 65 | ||
XM_018095385.1 | XP_017950874.1 | 8 - 9 | 1532 | 65 | ||
XM_018095386.1 | XP_017950875.1 | 6 - 7 | 1864 | 65 | ||
GenBank mRNA | CR926390.2 | - | 7 - 8 | 1653 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Xenopus tropicalis Probe spans exons |