Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: phgdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015929.2 | NP_001015929.1 | 4 - 5 | 488 | 91 | |
GenBank mRNA | BC118688.1 | - | 4 - 5 | 497 | 91 | |
CR926379.2 | - | 4 - 5 | 488 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |