Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pycr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015942.2 | NP_001015942.1 | 5 - 6 | 975 | 95 | |
GenBank mRNA | CR848490.2 | - | 5 - 6 | 975 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Xenopus tropicalis Probe spans exons |