Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sugp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015943.2 | NP_001015943.1 | 8 - 9 | 1165 | 67 | |
GenBank mRNA | BC161578.1 | - | 8 - 9 | 1166 | 67 | |
CR848489.2 | - | 8 - 9 | 1165 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |