Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tpte2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015951.2 | NP_001015951.1 | 7 - 8 | 682 | 94 | |
XM_018091231.1 | XP_017946720.1 | 7 - 8 | 940 | 94 | ||
XM_018091232.1 | XP_017946721.1 | 8 - 9 | 1004 | 94 | ||
XM_018091233.1 | XP_017946722.1 | 8 - 9 | 1577 | 94 | ||
XM_018091234.1 | XP_017946723.1 | 7 - 8 | 655 | 94 | ||
GenBank mRNA | BC121583.1 | - | 2 - 3 | 153 | 94 | |
BC154861.1 | - | 4 - 5 | 422 | 94 | ||
CR848475.2 | - | 7 - 8 | 682 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |