Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mien1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015996.2 | NP_001015996.1 | 3 - 4 | 205 | 89 | |
GenBank mRNA | BC122005.1 | - | 3 - 4 | 204 | 89 | |
CR848294.2 | - | 3 - 4 | 205 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |