Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: reep6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016051.2 | NP_001016051.1 | 3 - 4 | 380 | 107 | |
XM_012961953.2 | XP_012817407.1 | 3 - 4 | 432 | 107 | ||
GenBank mRNA | BC122013.1 | - | 3 - 4 | 381 | 107 | |
CR762344.2 | - | 3 - 4 | 380 | 107 | ||
CT025233.2 | - | 3 - 4 | 409 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |