Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vps25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016078.2 | NP_001016078.1 | 5 - 6 | 423 | 83 | |
GenBank mRNA | BC121525.1 | - | 5 - 6 | 425 | 83 | |
CR762222.2 | - | 5 - 6 | 423 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |