Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: akirin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016080.2 | NP_001016080.1 | 2 - 3 | 438 | 76 | |
GenBank mRNA | BC090594.1 | - | 2 - 3 | 467 | 76 | |
CR762217.2 | - | 2 - 3 | 438 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |