Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: c7orf25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016099.2 | NP_001016099.1 | 1 | 1148 | 87 | |
XM_012964677.1 | XP_012820131.1 | 2 | 1565 | 87 | ||
GenBank mRNA | BC091034.1 | - | 2 | 1186 | 87 | |
CR762149.2 | - | 1 | 1148 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Xenopus tropicalis Both primers and probe map within a single exon |