Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: arl10b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016111.2 | NP_001016111.1 | 4 - 5 | 492 | 102 | |
GenBank mRNA | BC155533.1 | - | 4 - 5 | 527 | 102 | |
CR762112.2 | - | 4 - 5 | 492 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |