Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: metap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016115.2 | NP_001016115.1 | 3 - 4 | 345 | 106 | |
GenBank mRNA | BC161036.1 | - | 3 - 4 | 341 | 106 | |
CR762101.2 | - | 3 - 4 | 345 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |