Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: faap24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016122.2 | NP_001016122.1 | 1 - 2 | 133 | 91 | |
XM_012961062.1 | XP_012816516.1 | 2 - 3 | 91 | |||
GenBank mRNA | CR762074.2 | - | 1 - 2 | 133 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Xenopus tropicalis Probe spans exons |