Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mrpl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016126.2 | NP_001016126.1 | 2 - 3 | 167 | 108 | |
GenBank mRNA | BC098087.1 | - | 2 - 3 | 142 | 108 | |
BC158989.1 | - | 2 - 3 | 145 | 108 | ||
BC170718.1 | - | 2 - 3 | 167 | 108 | ||
BC170720.1 | - | 2 - 3 | 167 | 108 | ||
CR762063.2 | - | 2 - 3 | 167 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |