Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ube2w | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016132.2 | NP_001016132.1 | 2 - 3 | 152 | 79 | |
XM_012964678.2 | XP_012820132.1 | 2 - 3 | 194 | 79 | ||
XM_012964679.2 | XP_012820133.1 | 2 - 3 | 177 | 79 | ||
XM_012964681.2 | XP_012820135.1 | 2 - 3 | 178 | 79 | ||
XM_018094644.1 | XP_017950133.1 | 2 - 3 | 267 | 79 | ||
GenBank mRNA | BC157276.1 | - | 2 - 3 | 221 | 79 | |
CR762043.2 | - | 2 - 3 | 152 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |