Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: serbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016153.4 | NP_001016153.1 | 3 - 4 | 778 | 70 | |
GenBank mRNA | CR761958.2 | - | 3 - 4 | 676 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Xenopus tropicalis Probe spans exons |