Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: elovl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016159.2 | NP_001016159.1 | 5 - 6 | 550 | 103 | |
GenBank mRNA | BC135162.1 | - | 5 - 6 | 550 | 103 | |
CR761931.2 | - | 5 - 6 | 550 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |