Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sap18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016166.2 | NP_001016166.1 | 3 - 4 | 379 | 85 | |
GenBank mRNA | BC080173.1 | - | 3 - 4 | 381 | 85 | |
BC135392.1 | - | 3 - 4 | 363 | 85 | ||
CR760055.2 | - | 3 - 4 | 323 | 85 | ||
CR761908.2 | - | 3 - 4 | 379 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |