Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rpl27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016180.2 | NP_001016180.1 | 2 - 3 | 257 | 99 | |
XM_012971761.2 | XP_012827215.1 | 3 - 4 | 362 | 99 | ||
GenBank mRNA | BC080487.1 | - | 3 - 4 | 290 | 99 | |
CR761841.2 | - | 2 - 3 | 257 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |