Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tor1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016237.2 | NP_001016237.1 | 4 - 5 | 828 | 93 | |
GenBank mRNA | BC161275.1 | - | 4 - 5 | 763 | 93 | |
BC170808.1 | - | 4 - 5 | 813 | 93 | ||
BC171143.1 | - | 4 - 5 | 813 | 93 | ||
CR926267.2 | - | 4 - 5 | 828 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |