Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: smim19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016254.3 | NP_001016254.2 | 2 - 3 | 218 | 83 | |
NM_001244887.1 | NP_001231816.1 | 2 - 3 | 209 | 83 | ||
XM_012971763.2 | XP_012827217.1 | 2 - 3 | 277 | 83 | ||
GenBank mRNA | BC135856.1 | - | 2 - 3 | 151 | 83 | |
CN120506.1 | - | 2 - 3 | 218 | 83 | ||
CR421163.1 | - | 2 - 3 | 209 | 83 | ||
CR761744.2 | - | 1 - 2 | 138 | 83 | ||
CX373428.2 | - | 2 - 3 | 151 | 83 | ||
CX514979.2 | - | 2 - 3 | 193 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |